Assay Details
Target Gene Details
Entrez Gene ID: | 23583 |
Gene Name: | single-strand-selective monofunctional uracil-DNA glycosylase 1 |
Gene Aliases: |
FDG, HMUDG, UNG3 |
Location: |
Chr.12:54171815-54188994 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832419 | Chr.12:54064043 - 54259156 on Build GRCh38 | Loss |
|
| nsv509467 | Chr.12:54173617 - 54255598 on Build GRCh38 | Insertion |
|
| nsv832418 | Chr.12:54016556 - 54186660 on Build GRCh38 | Loss |
|
| nsv832420 | Chr.12:54137272 - 54296797 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map