Assay Details
Target Gene Details
Entrez Gene ID: | 8549 |
Gene Name: | leucine rich repeat containing G protein-coupled receptor 5 |
Gene Aliases: |
FEX, GPR49, GPR67, GRP49, HG38 |
Location: |
Chr.12:71439770-71586310 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LGR5 | NM_001277226.1 | NP_001264155.1 | ||
| NM_001277227.1 | NP_001264156.1 | |||
| NM_003667.3 | NP_003658.1 | |||
| NR_110596.1 | ||||
| AB105190.1 | ||||
| AK294985.1 | ||||
| AK315133.1 | ||||
| DC323352.1 | ||||
| DR003044.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2762990 | Chr.12:71428621 - 71501180 on Build GRCh38 | Gain |
|
| esv2761748 | Chr.12:71341747 - 71472405 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map