Assay Details
Target Gene Details
Entrez Gene ID: | 2842 |
Gene Name: | G protein-coupled receptor 19 |
Gene Aliases: |
- |
Location: |
Chr.12:12659691-12716305 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR19 | NM_006143.2 | NP_006134.1 | ||
| XM_011520623.2 | XP_011518925.1 | |||
| XM_011520624.2 | XP_011518926.1 | |||
| XM_011520625.2 | XP_011518927.1 | |||
| XM_017019212.1 | XP_016874701.1 | |||
| XM_017019213.1 | XP_016874702.1 | |||
| XM_017019214.1 | XP_016874703.1 | |||
| XM_017019215.1 | XP_016874704.1 | |||
| XM_017019216.1 | XP_016874705.1 | |||
| AK074729.1 | ||||
| DB022290.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2745567 | Chr.12:12246086 - 13092998 on Build GRCh38 | Deletion |
|
| nsv509456 | Chr.12:12645094 - 12681838 on Build GRCh38 | Insertion |
|
| nsv1046355 | Chr.12:11501836 - 12890185 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73281125] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map