Assay Details
Target Gene Details
Entrez Gene ID: | 84324 |
Gene Name: | SAP domain containing ribonucleoprotein |
Gene Aliases: |
CIP29, HCC1, HSPC316, THO1 |
Location: |
Chr.12:55752463-55817756 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SARNP | NM_033082.3 | NP_149073.1 | ||
| NR_026722.1 | ||||
| NR_026723.1 | ||||
| AF161434.1 | AAF28994.1 | |||
| AF486281.1 | AAM09686.1 | |||
| AK290508.1 | ||||
| BC007099.1 | AAH07099.1 | |||
| BC093051.1 | AAH93051.1 | |||
| BI520421.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832423 | Chr.12:55687038 - 55859914 on Build GRCh38 | Gain |
|
| nsv559004 | Chr.12:55704254 - 55777109 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map