Assay Details
Target Gene Details
Entrez Gene ID: | 4801 |
Gene Name: | nuclear transcription factor Y subunit beta |
Gene Aliases: |
CBF-A, CBF-B, HAP3, NF-YB |
Location: |
Chr.12:104117080-104138262 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NFYB | NM_006166.3 | NP_006157.1 | ||
| XM_005268908.4 | XP_005268965.1 | |||
| XM_005268909.4 | XP_005268966.1 | |||
| XM_005268910.4 | XP_005268967.1 | |||
| AK291934.1 | ||||
| AK311706.1 | 3 | 491 | ||
| BC005316.1 | AAH05316.1 | |||
| BC005317.1 | AAH05317.1 | |||
| BC007035.1 | AAH07035.1 | |||
| BG776259.1 | ||||
| X59710.1 | CAA42230.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3580446 | Chr.12:104129444 - 104138892 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map