Assay Details
Target Gene Details
Entrez Gene ID: | 8655 |
Gene Name: | dynein light chain LC8-type 1 |
Gene Aliases: |
DLC1, DLC8, DNCL1, DNCLC1, LC8, LC8a, PIN, hdlc1 |
Location: |
Chr.12:120469857-120498495 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DYNLL1 | NM_001037494.1 | NP_001032583.1 | ||
| BG474448.1 | ||||
| CB992195.1 |
Target Gene Details
Entrez Gene ID: | 100506668 |
Gene Name: | negative regulator of antiviral response (non-protein coding) |
Gene Aliases: |
DYNLL1-AS1, DYNLL1AS1 |
Location: |
Chr.12:120490338-120495946 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NRAV | NR_038854.1 | 2 | 713 | |
| AK026783.1 | 2 | 692 | ||
| BX537644.1 | 2 | 715 | ||
| KF311770.2 | 2 | 676 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509482 | Chr.12:120435117 - 120495113 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map