Assay Details
Target Gene Details
Entrez Gene ID: | 488 |
Gene Name: | ATPase sarcoplasmic/endoplasmic reticulum Ca2+ transporting 2 |
Gene Aliases: |
ATP2B, DAR, DD, SERCA2 |
Location: |
Chr.12:110281227-110351093 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP2A2 | NM_001681.3 | NP_001672.1 | ||
| NM_170665.3 | NP_733765.1 | |||
| XM_005253888.2 | XP_005253945.1 | |||
| XM_011538402.2 | XP_011536704.1 | |||
| BC035588.1 | AAH35588.1 | |||
| KU177929.1 | ||||
| KU177930.1 | ||||
| KU177931.1 | ||||
| M23114.1 | AAA53193.1 | |||
| M23115.1 | AAA53194.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1044428 | Chr.12:110227288 - 110387994 on Build GRCh38 | Gain |
|
| nsv560154 | Chr.12:110281203 - 110284827 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map