Assay Details
Target Gene Details
Entrez Gene ID: | 283316 |
Gene Name: | CD163 molecule like 1 |
Gene Aliases: |
CD163B, M160, SCARI2 |
Location: |
Chr.12:7320510-7444185 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CD163L1 | XM_011520616.1 | 19 | 5316 | XP_011518918.1 |
| XM_011520617.2 | 19 | 5421 | XP_011518919.1 | |
| XM_011520618.2 | 18 | 4740 | XP_011518920.1 | |
| XM_011520619.1 | 19 | 4932 | XP_011518921.1 | |
| XM_011520620.1 | 18 | 4886 | XP_011518922.1 | |
| XM_011520621.2 | 13 | 3425 | XP_011518923.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv594 | Chr.12:7329134 - 7363501 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map