Assay Details
Target Gene Details
Entrez Gene ID: | 8655 |
Gene Name: | dynein light chain LC8-type 1 |
Gene Aliases: |
DLC1, DLC8, DNCL1, DNCLC1, LC8, LC8a, PIN, hdlc1 |
Location: |
Chr.12:120469857-120498495 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DYNLL1 | NM_001037494.1 | NP_001032583.1 | ||
| NM_001037495.1 | NP_001032584.1 | |||
| NM_003746.2 | NP_003737.1 | |||
| AK309022.1 | 2 | 1147 | ||
| AK311790.1 | ||||
| BC100289.1 | AAI00290.1 | |||
| BC104244.1 | AAI04245.1 | |||
| BC104245.1 | AAI04246.1 | |||
| BG474448.1 | ||||
| BG705617.1 | ||||
| BM817665.1 | ||||
| CB992195.1 | ||||
| CD678447.1 | ||||
| CR407672.1 | CAG28600.1 | |||
| CR542128.1 | CAG46925.1 | |||
| U32944.1 | AAB04149.1 |
More Information
Set Membership: |
|

Genomic Map