Assay Details
Target Gene Details
Entrez Gene ID: | 204010 |
Gene Name: | ribosomal protein SA pseudogene 52 |
Gene Aliases: |
RPSA_17_1251 |
Location: |
Chr.12:65758020-65826974 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPSAP52 | NR_026825.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv275418 | Chr.12:65773874 - 65784968 on Build GRCh38 | Gain+Loss |
|
| dgv1521n100 | Chr.12:65772966 - 65810281 on Build GRCh38 | Gain |
|

Genomic Map