Assay Details
Target Gene Details
Entrez Gene ID: | 113251 |
Gene Name: | La ribonucleoprotein domain family member 4 |
Gene Aliases: |
PP13296 |
Location: |
Chr.12:50400809-50480005 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2761738 | Chr.12:50233509 - 50442644 on Build GRCh38 | Gain |
|
| dgv1498n100 | Chr.12:50240559 - 50407969 on Build GRCh38 | Gain |
|
| dgv1499n100 | Chr.12:50297256 - 50477759 on Build GRCh38 | Gain |
|
| nsv470291 | Chr.12:50399063 - 50465037 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map