Assay Details
Target Gene Details
Entrez Gene ID: | 84530 |
Gene Name: | serine/arginine repetitive matrix 4 |
Gene Aliases: |
KIAA1853, MU-MB-2.76, nSR100 |
Location: |
Chr.12:118981495-119163051 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRRM4 | NM_194286.3 | NP_919262.2 | ||
| AB058756.1 | ||||
| AK291361.1 | ||||
| BC152471.1 | ||||
| BC167162.1 | ||||
| BC167163.1 | ||||
| KC995185.1 | ||||
| KC995186.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs117813832] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map