Assay Details
Target Gene Details
Entrez Gene ID: | 101928471 |
Gene Name: | uncharacterized LOC101928471 |
Gene Aliases: |
- |
Location: |
Chr.12:24223271-24237965 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928471 | NR_120472.1 | |||
| NR_120473.1 | ||||
| BX106967.1 | ||||
| CR744081.1 | ||||
| DB076822.1 |
Target Gene Details
Entrez Gene ID: | 6660 |
Gene Name: | SRY-box 5 |
Gene Aliases: |
L-SOX5, L-SOX5B, L-SOX5F, LAMSHF |
Location: |
Chr.12:23529499-24562701 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SOX5 | NM_001261414.1 | NP_001248343.1 | ||
| NM_152989.3 | NP_694534.1 | |||
| XM_011520834.2 | XP_011519136.1 | |||
| XM_011520835.2 | XP_011519137.1 | |||
| XM_017019890.1 | XP_016875379.1 | |||
| XM_017019891.1 | XP_016875380.1 | |||
| XM_017019892.1 | XP_016875381.1 | |||
| XM_017019893.1 | XP_016875382.1 | |||
| XM_017019896.1 | XP_016875385.1 | |||
| XM_017019897.1 | XP_016875386.1 | |||
| XM_017019898.1 | XP_016875387.1 | |||
| XM_017019899.1 | XP_016875388.1 | |||
| AB081588.1 | BAC16233.1 | |||
| BC060773.1 | AAH60773.1 | |||
| JX570584.1 | ||||
| JX570585.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv261e214 | Chr.12:24216938 - 24224767 on Build GRCh38 | Loss |
|
| esv3628861 | Chr.12:24217468 - 24224767 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map