Assay Details
Target Gene Details
Entrez Gene ID: | 8835 |
Gene Name: | suppressor of cytokine signaling 2 |
Gene Aliases: |
CIS2, Cish2, SOCS-2, SSI-2, SSI2, STATI2 |
Location: |
Chr.12:93569822-93626236 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SOCS2 | XM_011538929.1 | XP_011537231.1 | ||
| XM_011538935.1 | XP_011537237.1 | |||
| XM_011538936.1 | XP_011537238.1 | |||
| AL161980.1 | 1 | 254 |
More Information
Set Membership: |
|

Genomic Map