Assay Details
Target Gene Details
Entrez Gene ID: | 94 |
Gene Name: | activin A receptor like type 1 |
Gene Aliases: |
ACVRLK1, ALK-1, ALK1, HHT, HHT2, ORW2, SKR3, TSR-I |
Location: |
Chr.12:51906913-51923361 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACVRL1 | NM_000020.2 | 10 | 3453 | NP_000011.2 |
| NM_001077401.1 | 9 | 3316 | NP_001070869.1 | |
| XM_005269235.2 | 11 | 3455 | XP_005269292.1 | |
| XM_011539008.1 | 7 | 2999 | XP_011537310.1 | |
| AK056725.1 | 1 | 1151 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv707 | Chr.12:51913177 - 51944329 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map