Assay Details
Target Gene Details
Entrez Gene ID: | 1368 |
Gene Name: | carboxypeptidase M |
Gene Aliases: |
- |
Location: |
Chr.12:68842200-68963240 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CPM |

Genomic Map