Assay Details
Target Gene Details
Entrez Gene ID: | 101928062 |
Gene Name: | long intergenic non-protein coding RNA 1481 |
Gene Aliases: |
- |
Location: |
Chr.12:70222190-70243360 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC01481 | NR_120460.1 | |||
| NR_120461.1 | ||||
| DA349921.1 | ||||
| DB462634.1 | ||||
| HY013176.1 | ||||
| HY048577.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv763 | Chr.12:70214096 - 70236518 on Build GRCh38 | Deletion |
|
| nsv559237 | Chr.12:70139848 - 70233539 on Build GRCh38 | Loss |
|
| esv3629882 | Chr.12:70219974 - 70226855 on Build GRCh38 | Loss |
|

Genomic Map