Assay Details
Target Gene Details
Entrez Gene ID: | 94 |
Gene Name: | activin A receptor like type 1 |
Gene Aliases: |
ACVRLK1, ALK-1, ALK1, HHT, HHT2, ORW2, SKR3, TSR-I |
Location: |
Chr.12:51906913-51923361 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACVRL1 | NM_000020.2 | NP_000011.2 | ||
| XM_005269235.2 | XP_005269292.1 | |||
| AK298493.1 | ||||
| DN999570.1 | ||||
| HM161905.1 | ||||
| HM161906.1 | ||||
| Z22533.1 | CAA80255.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv558895 | Chr.12:51872564 - 51920604 on Build GRCh38 | Loss |
|
| nsv527286 | Chr.12:51892992 - 51920604 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map