Assay Details
Target Gene Details
Entrez Gene ID: | 55711 |
Gene Name: | fatty acyl-CoA reductase 2 |
Gene Aliases: |
HEL-S-81, MLSTD1, SDR10E2 |
Location: |
Chr.12:29149003-29335616 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAR2 | NM_001271783.1 | NP_001258712.1 | ||
| NM_001271784.1 | NP_001258713.1 | |||
| NM_018099.4 | NP_060569.3 | |||
| XM_011520747.2 | XP_011519049.1 | |||
| XM_011520748.2 | XP_011519050.1 | |||
| XM_017019624.1 | XP_016875113.1 | |||
| XM_017019625.1 | XP_016875114.1 | |||
| AK001324.1 | BAA91625.1 | |||
| AK001927.1 | BAA91983.1 | |||
| AK027756.1 | BAB55347.1 | |||
| AK129857.1 | ||||
| AK314670.1 | ||||
| AL136843.1 | CAB66777.1 | |||
| BC022267.1 | AAH22267.1 | |||
| GQ472211.1 |
Target Gene Details
Entrez Gene ID: | 100506606 |
Gene Name: | uncharacterized LOC100506606 |
Gene Aliases: |
- |
Location: |
Chr.12:29280415-29317848 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100506606 | NR_103860.1 | |||
| AK090590.1 |
More Information
Set Membership: |
|

Genomic Map