Assay Details
Target Gene Details
Entrez Gene ID: | 55110 |
Gene Name: | mago homolog B, exon junction complex core component |
Gene Aliases: |
MGN2, mago, magoh |
Location: |
Chr.12:10604190-10613609 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAGOHB | NM_001300739.2 | NP_001287668.1 | ||
| NM_001319985.1 | NP_001306914.1 | |||
| NM_018048.4 | NP_060518.1 | |||
| NR_135120.1 | ||||
| NR_135121.1 | ||||
| XM_005253402.1 | XP_005253459.1 | |||
| AF165518.1 | AAF86648.1 | |||
| AK001154.1 | BAA91522.1 | |||
| AK022720.1 | BAB14202.1 | |||
| AK096106.1 | ||||
| BC010905.1 | AAH10905.1 | |||
| BF667243.1 | ||||
| BG533740.1 | ||||
| BG539870.1 | ||||
| BU933036.1 | ||||
| CD357202.1 | ||||
| DB340467.1 | ||||
| KF051004.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832329 | Chr.12:10524495 - 10693347 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map