Assay Details
Target Gene Details
Entrez Gene ID: | 4848 |
Gene Name: | CCR4-NOT transcription complex subunit 2 |
Gene Aliases: |
CDC36, HSPC131, NOT2, NOT2H |
Location: |
Chr.12:70242993-70354993 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3629884 | Chr.12:70233284 - 70252849 on Build GRCh38 | Gain |
|
| nsv1048888 | Chr.12:70245128 - 70281639 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map