Assay Details
Target Gene Details
Entrez Gene ID: | 8091 |
Gene Name: | high mobility group AT-hook 2 |
Gene Aliases: |
BABL, HMGI-C, HMGIC, LIPO, STQTL9 |
Location: |
Chr.12:65824460-65966291 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HMGA2 | NM_001300918.1 | NP_001287847.1 | ||
| NM_001300919.1 | NP_001287848.1 | |||
| NM_003483.4 | NP_003474.1 | |||
| NM_003484.1 | NP_003475.1 | |||
| AB209853.1 | BAD93090.1 | |||
| AF533652.1 | ||||
| AF533654.1 | AAO48742.1 | |||
| AI290477.1 | ||||
| AY601863.1 | ||||
| AY601864.1 | ||||
| AY601865.1 | ||||
| AY601866.1 | ||||
| AY601867.1 | ||||
| CA422738.1 | ||||
| U28749.1 | AAA68613.1 | |||
| X92518.1 | CAA63295.1 | |||
| Z31595.1 | CAA83472.1 |
More Information
Set Membership: |
|

Genomic Map