Assay Details
Target Gene Details
Entrez Gene ID: | 8738 |
Gene Name: | CASP2 and RIPK1 domain containing adaptor with death domain |
Gene Aliases: |
MRT34, RAIDD |
Location: |
Chr.12:93677342-93894840 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CRADD | NM_001320099.1 | NP_001307028.1 | ||
NM_001320100.1 | NP_001307029.1 | |||
NM_001320101.1 | NP_001307030.1 | |||
NM_003805.4 | NP_003796.1 | |||
NR_135147.1 | ||||
XM_017020142.1 | XP_016875631.1 | |||
XM_017020143.1 | XP_016875632.1 | |||
XM_017020144.1 | XP_016875633.1 | |||
XM_017020145.1 | XP_016875634.1 | |||
AB451280.1 | ||||
AB451412.1 | ||||
AI417529.1 | ||||
AI791547.1 | ||||
AK294986.1 | ||||
AK296225.1 | ||||
AK314861.1 | ||||
BC017042.1 | AAH17042.1 | |||
BC037905.1 | AAH37905.1 | |||
BT009837.1 | AAP88839.1 | |||
CR407643.1 | CAG28571.1 | |||
HY113325.1 | ||||
U79115.1 | AAB42217.1 | |||
U84388.1 | AAC50952.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
