Assay Details
Target Gene Details
Entrez Gene ID: | 100507533 |
Gene Name: | SOX21 antisense RNA 1 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.13:94712716-94715945 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SOX21-AS1 | NR_046514.1 | 2 | 2512 | |
| AK055459.1 | 2 | 2514 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832686 | Chr.13:94554488 - 94748738 on Build GRCh38 | Gain |
|
| esv34004 | Chr.13:94254283 - 94740390 on Build GRCh38 | Loss |
|
| nsv826750 | Chr.13:94706248 - 94715816 on Build GRCh38 | Gain |
|

Genomic Map