Assay Details
Target Gene Details
Entrez Gene ID: | 9617 |
Gene Name: | mitochondrial translational release factor 1 |
Gene Aliases: |
MRF1, MTTRF1, RF1 |
Location: |
Chr.13:41216369-41311794 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
More Information
Set Membership: |
|

Genomic Map