Assay Details
Target Gene Details
Entrez Gene ID: | 8848 |
Gene Name: | TSC22 domain family member 1 |
Gene Aliases: |
Ptg-2, TGFB1I4, TSC22 |
Location: |
Chr.13:44432143-44576565 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSC22D1 | NM_001243797.1 | 3 | 2209 | NP_001230726.1 |
| NM_001243798.1 | 3 | 2214 | NP_001230727.1 | |
| NM_001243799.1 | 2 | 3363 | NP_001230728.1 | |
| NM_006022.3 | 3 | 2551 | NP_006013.1 | |
| NM_183422.3 | 3 | 5533 | NP_904358.2 |
More Information
Set Membership: |
|

Genomic Map