Assay Details
Target Gene Details
Entrez Gene ID: | 100533467 |
Gene Name: | BIVM-ERCC5 readthrough |
Gene Aliases: |
ERCC5-202 |
Location: |
Chr.13:102807146-102876001 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BIVM-ERCC5 | NM_001204425.1 | NP_001191354.1 | ||
| AB209312.1 | BAD92549.1 | |||
| LM994822.1 |
Target Gene Details
Entrez Gene ID: | 2073 |
Gene Name: | ERCC excision repair 5, endonuclease |
Gene Aliases: |
COFS3, ERCC5-201, ERCM2, UVDR, XPG, XPGC |
Location: |
Chr.13:102845841-102876001 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ERCC5 | NM_000123.3 | NP_000114.2 | ||
| AF462447.1 | AAP97715.1 | |||
| AK294708.1 | ||||
| AK299758.1 | ||||
| AK303558.1 | ||||
| BC031522.2 | AAH31522.1 | |||
| BX647399.1 | ||||
| D16305.1 | BAA03812.1 | |||
| DA195001.1 | ||||
| L20046.1 | AAC37533.1 | |||
| X69978.1 | CAA49598.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv562981 | Chr.13:102820720 - 103931125 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map