Assay Details
Target Gene Details
Entrez Gene ID: | 2581 |
Gene Name: | galactosylceramidase |
Gene Aliases: |
- |
Location: |
Chr.14:87933014-87993665 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GALC | NM_000153.3 | NP_000144.2 | ||
| NM_001201401.1 | NP_001188330.1 | |||
| NM_001201402.1 | NP_001188331.1 | |||
| XM_011536618.2 | XP_011534920.1 | |||
| AK294040.1 | ||||
| AK296530.1 | ||||
| AK302519.1 | ||||
| AK302683.1 | ||||
| AK302956.1 | ||||
| BC036518.2 | AAH36518.1 | |||
| BU675449.1 | ||||
| D25283.1 | BAA04971.1 | |||
| D25284.1 | BAA04972.1 | |||
| DB366392.1 | ||||
| L23116.1 | AAA16645.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs74796499] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map