Assay Details
Target Gene Details
Entrez Gene ID: | 3183 |
Gene Name: | heterogeneous nuclear ribonucleoprotein C (C1/C2) |
Gene Aliases: |
C1, C2, HNRNP, HNRPC, SNRPC |
Location: |
Chr.14:21209136-21269479 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3633707 | Chr.14:21222351 - 21227315 on Build GRCh38 | Loss |
|
| esv2656744 | Chr.14:21222370 - 21227289 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map