Assay Details
Target Gene Details
Entrez Gene ID: | 394 |
Gene Name: | Rho GTPase activating protein 5 |
Gene Aliases: |
GFI2, RhoGAP5, p190-B, p190BRhoGAP |
Location: |
Chr.14:32076152-32159728 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGAP5 | NM_001030055.1 | NP_001025226.1 | ||
| NM_001173.2 | NP_001164.2 | |||
| XM_005267635.3 | XP_005267692.1 | |||
| XM_005267636.3 | XP_005267693.1 | |||
| XM_017021287.1 | XP_016876776.1 | |||
| XM_017021288.1 | XP_016876777.1 | |||
| BC050059.1 | ||||
| BC075799.1 | AAH75799.1 | |||
| BC129928.1 | ||||
| BC129929.1 | ||||
| U17032.1 | AAA95963.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv564182 | Chr.14:31641313 - 32115131 on Build GRCh38 | Gain |
|
| nsv1046526 | Chr.14:31641331 - 32098220 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs116370397] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map