Assay Details
Target Gene Details
Entrez Gene ID: | 51635 |
Gene Name: | dehydrogenase/reductase 7 |
Gene Aliases: |
CGI-86, SDR34C1, retDSR4, retSDR4 |
Location: |
Chr.14:60144119-60169864 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DHRS7 | NM_001322280.1 | NP_001309209.1 | ||
| NM_001322281.1 | NP_001309210.1 | |||
| NM_001322282.1 | NP_001309211.1 | |||
| NM_016029.3 | NP_057113.1 | |||
| AF151844.1 | AAD34081.1 | |||
| AK313285.1 | ||||
| AL117420.1 | CAH56402.1 | |||
| AY359031.1 | AAQ89390.1 | |||
| BC000637.2 | AAH00637.1 | |||
| BC007337.2 | AAH07337.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1044390 | Chr.14:60109151 - 60168783 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map