Assay Details
Target Gene Details
Entrez Gene ID: | 145482 |
Gene Name: | prostaglandin reductase 2 |
Gene Aliases: |
HEL-S-298, PGR2, ZADH1 |
Location: |
Chr.14:73851831-73885465 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTGR2 | NM_001146154.1 | NP_001139626.1 | ||
| NM_001146155.1 | NP_001139627.1 | |||
| NM_152444.2 | NP_689657.1 | |||
| AK096410.1 | BAC04781.1 | |||
| AK309188.1 | ||||
| AK311280.1 | ||||
| AY346133.1 | AAR05101.1 | |||
| AY424308.1 | AAR82927.1 | |||
| BC059364.1 | AAH59364.1 | |||
| BP357520.1 | ||||
| BX641118.1 | CAE46055.1 | |||
| DR158023.1 | ||||
| FJ224334.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1053281 | Chr.14:73518500 - 74050155 on Build GRCh38 | Gain |
|
| nsv1035460 | Chr.14:72854572 - 73897173 on Build GRCh38 | Gain |
|
| nsv976844 | Chr.14:73850066 - 73884634 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map