Assay Details
Target Gene Details
Entrez Gene ID: | 79944 |
Gene Name: | L-2-hydroxyglutarate dehydrogenase |
Gene Aliases: |
C14orf160, L2HGA |
Location: |
Chr.14:50242434-50312235 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| L2HGDH | NM_024884.2 | NP_079160.1 | ||
| XM_005268075.4 | XP_005268132.1 | |||
| XM_011537166.2 | XP_011535468.1 | |||
| XM_011537167.2 | XP_011535469.1 | |||
| XM_011537168.2 | XP_011535470.1 | |||
| XM_017021655.1 | XP_016877144.1 | |||
| XM_017021656.1 | XP_016877145.1 | |||
| XM_017021657.1 | XP_016877146.1 | |||
| AK022680.1 | BAB14174.1 | |||
| AK292627.1 | ||||
| AY757363.1 | AAV52330.1 | |||
| BC006117.1 | AAH06117.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv564835 | Chr.14:50024482 - 50439753 on Build GRCh38 | Gain |
|
| esv28926 | Chr.14:49426970 - 50475809 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map