Assay Details
Target Gene Details
Entrez Gene ID: | 51062 |
Gene Name: | atlastin GTPase 1 |
Gene Aliases: |
AD-FSP, FSP1, GBP3, HSN1D, SPG3, SPG3A, atlastin1 |
Location: |
Chr.14:50533082-50633068 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATL1 | NM_001127713.1 | NP_001121185.1 | ||
| NM_015915.4 | NP_056999.2 | |||
| NM_181598.3 | NP_853629.2 | |||
| AF444143.1 | AAL37898.1 | |||
| AK223436.1 | BAD97156.1 | |||
| AK290185.1 | ||||
| AK312518.1 | ||||
| AL833591.1 | CAH10392.1 | |||
| AY032844.1 | AAK51160.1 | |||
| BC010708.2 | AAH10708.2 | |||
| BG704589.1 | ||||
| BI601372.1 | ||||
| DA088230.1 | ||||
| DA397302.1 | ||||
| KU178621.1 | ||||
| KU178622.1 | ||||
| KU178623.1 | ||||
| KU178624.1 |
Target Gene Details
Entrez Gene ID: | 11183 |
Gene Name: | mitogen-activated protein kinase kinase kinase kinase 5 |
Gene Aliases: |
GCKR, KHS, KHS1, MAPKKKK5 |
Location: |
Chr.14:50418493-50561126 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAP4K5 | XM_011536378.2 | XP_011534680.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1039493 | Chr.14:50476979 - 50687775 on Build GRCh38 | Gain |
|
| esv3581192 | Chr.14:50546798 - 50708585 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map