Assay Details
Target Gene Details
Entrez Gene ID: | 1033 |
Gene Name: | cyclin dependent kinase inhibitor 3 |
Gene Aliases: |
CDI1, CIP2, KAP, KAP1 |
Location: |
Chr.14:54396939-54420218 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDKN3 | NM_001130851.1 | NP_001124323.1 | ||
| NM_005192.3 | NP_005183.2 | |||
| XM_011536353.2 | XP_011534655.1 | |||
| AK098653.1 | ||||
| KP966095.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832801 | Chr.14:54255297 - 54420055 on Build GRCh38 | Gain |
|
| nsv952844 | Chr.14:54391983 - 54398182 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map