Assay Details
Target Gene Details
Entrez Gene ID: | 30001 |
Gene Name: | endoplasmic reticulum oxidoreductase 1 alpha |
Gene Aliases: |
ERO1-L, ERO1-L-alpha, ERO1-alpha, ERO1L, ERO1LA, Ero1alpha |
Location: |
Chr.14:52639915-52695931 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ERO1A | NM_014584.2 | NP_055399.1 | ||
| XM_011536684.2 | XP_011534986.1 | |||
| AF081886.1 | AAF35260.1 | |||
| AF123887.1 | AAF06104.1 | |||
| AK292839.1 | ||||
| AY358463.1 | AAQ88828.1 | |||
| BC008674.1 | AAH08674.1 | |||
| BC012941.1 | AAH12941.1 |
More Information
Set Membership: |
|

Genomic Map