Assay Details
Target Gene Details
Entrez Gene ID: | 54331 |
Gene Name: | G protein subunit gamma 2 |
Gene Aliases: |
- |
Location: |
Chr.14:51847131-51969800 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GNG2 | NM_001243773.1 | NP_001230702.1 | ||
| NM_001243774.1 | NP_001230703.1 | |||
| NM_053064.4 | NP_444292.1 | |||
| XM_006720173.2 | XP_006720236.1 | |||
| XM_011536846.2 | XP_011535148.1 | |||
| XM_017021376.1 | XP_016876865.1 | |||
| XM_017021377.1 | XP_016876866.1 | |||
| AK001024.1 | ||||
| AK092371.1 | ||||
| AK307171.1 | ||||
| AK311334.1 | ||||
| AL832878.1 | CAI46198.1 | |||
| BC020774.1 | AAH20774.1 | |||
| BE245271.1 | ||||
| DA226565.1 | ||||
| DA322879.1 | ||||
| DA785895.1 | ||||
| DC318652.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv456317 | Chr.14:51803013 - 51867871 on Build GRCh38 | Loss |
|
| nsv952840 | Chr.14:51846583 - 51949982 on Build GRCh38 | Duplication |
|
More Information
Additional Information:
For this assay, SNP(s) [rs72678121] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map