Assay Details
Target Gene Details
Entrez Gene ID: | 112849 |
Gene Name: | trans-L-3-hydroxyproline dehydratase |
Gene Aliases: |
C14orf149 |
Location: |
Chr.14:59465545-59484438 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| L3HYPDH | NM_144581.1 | NP_653182.1 | ||
| XM_005267298.3 | XP_005267355.1 | |||
| XM_011536386.2 | XP_011534688.1 | |||
| XM_011536387.1 | XP_011534689.1 | |||
| XM_011536388.1 | XP_011534690.1 | |||
| AK058165.1 | BAB71696.1 | |||
| BC012131.1 | AAH12131.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832810 | Chr.14:59317451 - 59478189 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map