Assay Details
Target Gene Details
Entrez Gene ID: | 8814 |
Gene Name: | cyclin dependent kinase like 1 |
Gene Aliases: |
KKIALRE, P42 |
Location: |
Chr.14:50329404-50397404 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDKL1 | NM_001282236.1 | NP_001269165.1 | ||
| NM_004196.4 | NP_004187.2 | |||
| XM_005268157.2 | XP_005268214.1 | |||
| XM_005268159.2 | XP_005268216.1 | |||
| XM_005268160.4 | XP_005268217.1 | |||
| XM_011537276.1 | XP_011535578.1 | |||
| XM_017021729.1 | XP_016877218.1 | |||
| XM_017021730.1 | XP_016877219.1 | |||
| XM_017021731.1 | XP_016877220.1 | |||
| XM_017021732.1 | XP_016877221.1 | |||
| AA780354.1 | ||||
| AF390028.1 | AAL58838.1 | |||
| AK308732.1 | ||||
| BC104977.1 | AAI04978.1 | |||
| X66358.1 | CAA47002.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv564835 | Chr.14:50024482 - 50439753 on Build GRCh38 | Gain |
|
| nsv832793 | Chr.14:50294245 - 50470857 on Build GRCh38 | Loss |
|
| nsv1052059 | Chr.14:50291535 - 50364621 on Build GRCh38 | Loss |
|
| esv28926 | Chr.14:49426970 - 50475809 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map