Assay Details
Target Gene Details
Entrez Gene ID: | 123016 |
Gene Name: | tetratricopeptide repeat domain 8 |
Gene Aliases: |
BBS8, RP51 |
Location: |
Chr.14:88824153-88877996 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832850 | Chr.14:88814253 - 88871220 on Build GRCh38 | Gain+Loss |
|
| dgv1955n100 | Chr.14:88712120 - 88999575 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map