Assay Details
Target Gene Details
Entrez Gene ID: | 9787 |
Gene Name: | DLG associated protein 5 |
Gene Aliases: |
DLG7, HURP |
Location: |
Chr.14:55148116-55191678 on Build GRCh38 |
Assay Gene Location: | Within Intron 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DLGAP5 | NM_001146015.1 | NP_001139487.1 | ||
| NM_014750.4 | NP_055565.3 | |||
| XM_017021840.1 | XP_016877329.1 | |||
| AB076695.1 | BAB97376.1 | |||
| AI174844.1 | ||||
| AK291847.1 | ||||
| AK297480.1 | ||||
| AK299338.1 | ||||
| BC010658.1 | AAH10658.2 | |||
| BC016276.2 | AAH16276.2 | |||
| BT007344.1 | AAP36008.1 | |||
| BX248255.1 | CAD62583.1 | |||
| D13633.3 |
More Information
Set Membership: |
|

Genomic Map