Assay Details
Target Gene Details
Entrez Gene ID: | 1033 |
Gene Name: | cyclin dependent kinase inhibitor 3 |
Gene Aliases: |
CDI1, CIP2, KAP, KAP1 |
Location: |
Chr.14:54396939-54420218 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832801 | Chr.14:54255297 - 54420055 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map