Assay Details
Target Gene Details
Entrez Gene ID: | 79609 |
Gene Name: | valosin containing protein lysine methyltransferase |
Gene Aliases: |
C14orf138, METTL21D, VCP-KMT |
Location: |
Chr.14:50100456-50118343 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VCPKMT | XM_017021640.1 | 6 | 791 | XP_016877129.1 |
| XM_017021641.1 | 6 | 788 | XP_016877130.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv564835 | Chr.14:50024482 - 50439753 on Build GRCh38 | Gain |
|
| esv28926 | Chr.14:49426970 - 50475809 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map