Assay Details
Target Gene Details
Entrez Gene ID: | 80127 |
Gene Name: | basal body orientation factor 1 |
Gene Aliases: |
C14orf45, CCDC176 |
Location: |
Chr.14:74019350-74078351 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BBOF1 | NM_025057.2 | 7 | 850 | NP_079333.2 |
| XM_005268092.3 | 7 | 856 | XP_005268149.1 | |
| XM_005268097.3 | XP_005268154.1 | |||
| XM_005268098.4 | 5 | 489 | XP_005268155.1 | |
| XM_011537170.2 | 7 | 856 | XP_011535472.1 | |
| XM_011537171.2 | 7 | 856 | XP_011535473.1 | |
| XM_011537172.1 | 7 | 856 | XP_011535474.1 | |
| XM_011537174.2 | 7 | 856 | XP_011535476.1 | |
| XM_011537175.2 | 6 | 712 | XP_011535477.1 | |
| XM_011537176.2 | 8 | 840 | XP_011535478.1 | |
| XM_011537177.2 | 6 | 560 | XP_011535479.1 | |
| XM_011537178.2 | 6 | 531 | XP_011535480.1 | |
| XM_011537179.2 | 5 | 416 | XP_011535481.1 | |
| XM_017021659.1 | 7 | 856 | XP_016877148.1 | |
| XM_017021660.1 | 6 | 572 | XP_016877149.1 | |
| XM_017021661.1 | 6 | 624 | XP_016877150.1 | |
| XM_017021662.1 | 5 | 408 | XP_016877151.1 | |
| XM_017021663.1 | XP_016877152.1 | |||
| AK097531.1 | 4 | 420 | ||
| AL834487.1 | 2 | 125 | CAD39145.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1053281 | Chr.14:73518500 - 74050155 on Build GRCh38 | Gain |
|
| dgv1936n100 | Chr.14:73950133 - 74073431 on Build GRCh38 | Gain |
|
| dgv1938n100 | Chr.14:73984364 - 74075786 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114681409] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map