Assay Details
Target Gene Details
Entrez Gene ID: | 57452 |
Gene Name: | polypeptide N-acetylgalactosaminyltransferase 16 |
Gene Aliases: |
GALNACT16, GALNTL1, GalNAc-T16 |
Location: |
Chr.14:69259546-69379904 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GALNT16 | NM_001168368.1 | NP_001161840.1 | ||
| NM_020692.2 | NP_065743.2 | |||
| XM_011537005.1 | XP_011535307.1 | |||
| XM_011537006.2 | XP_011535308.1 | |||
| XM_011537007.2 | XP_011535309.1 | |||
| XM_017021498.1 | XP_016876987.1 | |||
| AB032956.1 | BAA86444.1 | |||
| AB078143.1 | BAD93178.1 | |||
| AJ505951.1 | CAD44534.1 | |||
| AK289745.1 | ||||
| AK299588.1 | ||||
| AK315848.1 | ||||
| BC036812.1 | ||||
| BC098578.1 | AAH98578.1 | |||
| BC122545.1 | ||||
| DA493083.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs74059925] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map