Assay Details
Target Gene Details
Entrez Gene ID: | 5693 |
Gene Name: | proteasome subunit beta 5 |
Gene Aliases: |
LMPX, MB1, X |
Location: |
Chr.14:23016543-23035220 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PSMB5 | NM_001130725.1 | NP_001124197.1 | ||
NM_001144932.2 | NP_001138404.1 | |||
NM_002797.4 | NP_002788.1 | |||
XM_005267871.2 | XP_005267928.1 | |||
AK225888.1 | ||||
AK300714.1 | ||||
AK311895.1 | ||||
BC057840.1 | AAH57840.1 | |||
BC107720.1 | AAI07721.1 | |||
BT006777.1 | AAP35423.1 | |||
BU539357.1 | ||||
BU632351.1 | ||||
BX248299.1 | CAD62626.1 | |||
BX538001.1 | CAD97956.1 | |||
CD048996.1 | ||||
S74378.1 | AAB33092.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
