Assay Details
Target Gene Details
Entrez Gene ID: | 5729 |
Gene Name: | prostaglandin D2 receptor |
Gene Aliases: |
AS1, ASRT1, DP, DP1, PTGDR1 |
Location: |
Chr.14:52267256-52280914 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTGDR | NM_000953.2 | NP_000944.1 | ||
| NM_001281469.1 | NP_001268398.1 | |||
| XM_005267891.3 | XP_005267948.1 | |||
| BC040968.1 | AAH40968.1 | |||
| CD013871.1 | ||||
| EF577397.1 | ||||
| U31099.1 | AAC50177.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1044604 | Chr.14:52131535 - 52289964 on Build GRCh38 | Gain |
|
| nsv832795 | Chr.14:52230515 - 52428270 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map