Assay Details
Target Gene Details
Entrez Gene ID: | 161159 |
Gene Name: | ACTN1 antisense RNA 1 |
Gene Aliases: |
C14orf84 |
Location: |
Chr.14:68979682-68987463 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACTN1-AS1 | NR_073422.1 | |||
| DB029126.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832823 | Chr.14:68922136 - 69100921 on Build GRCh38 | Loss |
|

Genomic Map