Assay Details
Target Gene Details
Entrez Gene ID: | 100996664 |
Gene Name: | uncharacterized LOC100996664 |
Gene Aliases: |
- |
Location: |
Chr.14:68625527-68628787 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100996664 | NR_135816.1 | 1 | 292 |
Target Gene Details
Entrez Gene ID: | 5890 |
Gene Name: | RAD51 paralog B |
Gene Aliases: |
R51H2, RAD51L1, REC2 |
Location: |
Chr.14:67819779-68683118 on Build GRCh38 |
Assay Gene Location: | Within Intron 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAD51B | NM_001321818.1 | NP_001308747.1 | ||
| BX248061.1 | CAD62357.1 |
More Information
Set Membership: |
|

Genomic Map