Assay Details
Target Gene Details
Entrez Gene ID: | 3091 |
Gene Name: | hypoxia inducible factor 1 alpha subunit |
Gene Aliases: |
HIF-1-alpha, HIF-1A, HIF-1alpha, HIF1, HIF1-ALPHA, MOP1, PASD8, bHLHe78 |
Location: |
Chr.14:61695401-61748259 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HIF1A | NM_001530.3 | NP_001521.1 | ||
| NM_181054.2 | NP_851397.1 | |||
| AB073325.1 | BAB70608.1 | |||
| AB500182.1 | ||||
| AB733094.1 | ||||
| AF207601.1 | AAF20139.1 | |||
| AF207602.1 | AAF20140.1 | |||
| AF304431.1 | AAG43026.1 | |||
| AK296876.1 | ||||
| AK299551.1 | ||||
| AK304315.1 | ||||
| AK304436.1 | ||||
| AK312397.1 | ||||
| AU123241.1 | ||||
| BC012527.2 | AAH12527.1 | |||
| BT009776.1 | AAP88778.1 | |||
| BX648795.1 | ||||
| DC344032.1 | ||||
| U22431.1 | AAC50152.1 | |||
| U29165.1 | AAC51210.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv564884 | Chr.14:61667038 - 61740839 on Build GRCh38 | Loss |
|
| dgv3721n54 | Chr.14:61682190 - 61740839 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map